missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nonanal-d18, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Specifications
Specifications
| Chemical Name or Material | Nonanal-d18 |
| CAS | 1466552-36-2 |
| Molecular Formula | C9 D18 O |
| Quantity | 0.25 g |
| Synonym | Nonanaldehyde-d18, Aldehyde C 09-d18, Aldehyde C 9-d18, NSC 5518-d18, Nonaldehyde-d18, Nonanoic Aldehyde-d18, Nonylaldehyde-d18, Nonylic Aldehyde-d18, Pelargonaldehyde-d18, Pelargonic Aldehyde-d18, n-Nonanal-d18, n-Nonylaldehyde-d18 |
| InChI Formula | InChI=1 S/C9H18O/c1-2-3-4-5-6-7-8-9-10/h9H,2-8H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9 D |
| SMILES | [2 H]C(=O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-octadecadeuteriononan-1-one |
| Molecular Weight (g/mol) | 160.35 |
| Formula Weight | 160.2487 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction