Learn More
Nitritex™ Non-sterile Cleanroom Gloves for Chemotherapy
Description
- Non-latex
- Various sizes
- Facilitate the safe handling of chemotherapy drugs
Specifications
Specifications
| Certifications/Compliance (Europe) | ISO 9001:2015 and (EU)2016/425. (EU)2016/425, EN420:2003 +A1:2009, EN374-1:2016, EN374-5:2016 and EN421:2010, EN455-2:2015, (EC)1935/2004 and (EC) 2023/2006. |
| Disposable | Single-use |
| Laboratory/Cleanroom Use | Yes |
| Latex Free | Yes |
| Material | Nitrile |
| Sterility | Non-sterile |
| Packaging Type | 100 pieces per sealed inner PE bag; one inner PE bag per sealed outer PE bag; 10 outer bags per lined carton (1000 pieces) |
| Quantity | 1000 Pk. |