Learn More
Laboratoires Anios™ Non-Autoclavable Instrubac
Click to view available options
Dimensions (L x W x H):
23 x 17 x 9 cm
Unit Size:
Each
Description
- Soaking bin
- Non-autoclavable
- Without slit
- Without tap
Specifications
Specifications
| Type | Soaking Bin |
| Capacity (Metric) | 2 L |
| Autoclavable | Not Autoclavable |
| Includes | Does not include slit or tap. |
| Dimensions (L x W x H) | 23 x 17 x 9 cm |
| Length (Metric) | 23 cm |
| Width (Metric) | 17 cm |
| Height (Metric) | 9 cm |