Learn More
Nocodazole, MP Biomedicals™
Product Code. 15997255 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15997255 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15997255 Supplier MP Biomedicals Supplier No. 0215240510
Used in Cell Biology to synchronize the cell division cycle, as well as in cell studies and cancer research when trying to understand the microspindles in the cell replication process.
  • Used in cell division & replication research, cancer research, microscopy, and flow cytometry
  • Nocodazole is usually used at a concentration of 40–100 ng/mL of culture medium for a duration of 12 to 18 hours
  • Prolonged arrest of cells in mitosis due to nocodazole treatment typically results in cell death by apoptosis
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg