Learn More
NLN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA598844
Additional Details : Weight : 0.01000kg
Description
This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.Specifications
NLN | |
Polyclonal | |
Unconjugated | |
Nln | |
4930472G13Rik; AGTBP; angiotensin binding protein; angiotensin-binding protein; C79345; endopeptidase 24.16; EP24.16; KIAA1226; MEP; microsomal endopeptidase; Mitochondrial oligopeptidase M; MOP; neurolysin; neurolysin (metallopeptidase M3 family); neurolysin, mitochondrial; Neurotensin endopeptidase; NLN | |
Rabbit | |
Protein G | |
RUO | |
57486 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
3.22 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9BYT8 | |
Nln | |
Recombinant Human Neurolysin, mitochondrial protein (109-241AA) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |