Learn More
NLN Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA588992
Additional Details : Weight : 0.01000kg
Description
Positive Samples: U-87MG, A-549, Rat liver; Cellular Location: Cytoplasm, Mitochondrion intermembrane space Immunogen sequence: DGQYYGYLWS EVFSMDMFYS CFKKEGIMNP EVGMKYRNLI LKPGGSLDGM DMLHNFLKRE PNQKAFLMSR GLHAP
This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.Specifications
NLN | |
Polyclonal | |
Unconjugated | |
Nln | |
4930472G13Rik, AGTBP, angiotensin binding protein, angiotensin-binding protein, C79345, endopeptidase 24.16, EP24.16, KIAA1226, MEP, microsomal endopeptidase, Mitochondrial oligopeptidase M, MOP, neurolysin, neurolysin (metallopeptidase M3 family), neurolysin, mitochondrial, Neurotensin endopeptidase, NLN | |
Rabbit | |
Affinity Chromatography | |
RUO | |
117041, 57486 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.91 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
P42676, Q9BYT8 | |
Nln | |
Recombinant fusion protein containing a sequence corresponding to amino acids 630-704 of human NLN (NP_0657771) | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |