Learn More
NKp46 Monoclonal Antibody (AKS1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA516573
Additional Details : Weight : 0.01000kg
Description
This product is a low-endotoxin, preservative-free formulation. Endotoxin level is <0.01EU/ug. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.
NKp46 (CD335, NCR1, ) is a cytotoxicity-activating receptor that may contribute to the increased efficiency of activated natural killer (NK) cells to mediate tumor cell lysis. NKp46 consists of two Ig-like domains assembled to leukocyte immunoglobulin-like (LIR) and killer inhibitory receptors (KIR).Specifications
NKp46 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q863H2 | |
NCR1 | |
Fusion protein consisting of the extracellular region of bovine CD335. | |
500 μg | |
Primary | |
Bison, Bovine | |
Antibody | |
IgG1 |
Flow Cytometry, Functional Assay, Immunocytochemistry, Western Blot | |
AKS1 | |
Unconjugated | |
NCR1 | |
1-Mar; activating receptor 1; Ar1; CD335; hNKp46; Kilr1; KILR-1; LY94; Ly-94; lymphocyte antigen 94; Lymphocyte antigen 94 homolog; lymphocyte antigen 94 homolog (activating NK-receptor; lymphocyte antigen 94 homolog (activating NK-receptor; NK-p46); mAR-1; MAR1 (mouse activating receptor 1); mNKp46; natural cytotoxicity triggering receptor 1; Natural killer cell p46-related protein; natural killer cell receptor; Ncr1; NK cell-activating receptor; NK receptor KILR-1; NKACTR; Nkp46; NK-p46; NK-p46); rAR-1; rat-activating receptor 1 | |
Mouse | |
Protein A | |
RUO | |
369024 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |