Learn More
Aalborg™ Nitrogen Direct Flow Tube 65mm Reference Scale
Product Code. p-8008829
Click to view available options
Airflow Rate:
12 L/min.
120 mL/min.
200 mL/min.
6 mL/min.
60 mL/min.
Unit Size:
Each
Product Code. 11862430
Brand: Aalborg™ FT01416STVN
Only null left
Add to basket
Add to basket
Aalborg™ Nitrogen Direct Flow Tube 65mm Reference Scale completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Type Nitrogen Direct Flow Tube
Width (Metric) 65 mm
Airflow Rate 12 L/min.