Learn More
Aalborg™ Nitrogen Direct Flow Tube 150mm Reference Scale
Product Code. 11748288
Change view
Click to view available options
Airflow Rate:
100 mL/min.
2 L/min.
200 mL/min.
27 L/min.
300 mL/min.
500 mL/min.
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Airflow Rate Unit Size
11748288 300 mL/min. Each
11718288 200 mL/min. Each
11788278 100 mL/min. Each
11768288 2 L/min. Each
11758288 500 mL/min. Each
11767739 27 L/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11748288 Supplier Aalborg™ Supplier No. FT06230CAVN
  • ONLINE EXCLUSIVE PRICE
    123.90€
    75.48€ / Pack of 1
    Save 48.42€
    39% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Aalborg™ Nitrogen Direct Flow Tube 150mm Reference Scale completes a flowmeter, which is used in fluid systems to indicate the rate of flow of the fluid.
  • Direct reading tubes are straightforward.
  • The measurement scale on each of these tubes reads actual flow at standard conditions in a choice of English or Metric units.
  • Obtaining actual flow rates with these tubes requires the use of a reference scale flow correlation table.
  • Reference scale tubes are useful when measuring flow rates for gases other than air and/or for non-standard conditions.
Specifications
Type Nitrogen Direct Flow Tube
Width (Metric) 150 mm
Airflow Rate 300 mL/min.