missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitritocobalamin (~90% by HPLC), TRC
Specifications
Specifications
| Chemical Name or Material | Nitritocobalamin |
| CAS | 20623-13-6 |
| Molecular Formula | C62H88CoN14O16P |
| Quantity | 10 mg |
| Synonym | Co-(nitrito-κO)-cobinamide Dihydrogen Phosphate (Ester) Inner Salt 3'-Ester with (5,6-Dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole-κN3), Nitrocobalamin, Vitamin B12c, Cobinamide','hydroxide nitrite (salt)','dihydrogen phosphate (ester)','inner salt','3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole, Cobinamide','nitrite (salt)','dihydrogen phosphate (ester)','inner salt','3'-ester with 5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole, Cobinamide','nitrite phosphate','3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole','inner salt |
| InChI Formula | InChI=1S/C62H90N13O14P.Co.HNO2/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;;2-1-3/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;(H,2,3)/q;+3;/p-3 |
| SMILES | -NC(CC1(C)C(CCC(N)=O)C2=N/C1=C(C)\C3=NC(C(C(CC(N)=O)C/4(CCC(NCC(OP(O[C@H]5[C@@H](O)[C@@H](N6C=[N+]([Co+]ON=O)C7=C6C=C(C)C(C)=C7)O[C@@H]5CO)([O-])=O)C)=O)C)[N-]C4=C(C)/C(C(CCC(N)=O)C/8(C)C)=NC8=C/2)(C)C(CC(N)=O)(C)C3CCC(N)=O)=O |
| Molecular Weight (g/mol) | 1375.35 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?