missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrilotriacetic Acid-d9, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Nitrilotriacetic Acid-d9 |
| CAS | 807630-34-8 |
| Molecular Formula | C6 D9 N O6 |
| Quantity | 0.1 g |
| Synonym | Glycine-1,2,2-d3, N,N-bis[(carboxy-d)methyl-d2]- (9 CI), N,N-Bis[(carboxy-d)methyl-d2]glycine-1,2,2-d3, Nitrilotriacetic Acid-d9, Nitrilotriacetic Acid-D9, NTA-d9, Deuterio 2-[bis(1,1-dideuterio-2-deuteriooxy-2-oxo-ethyl)amino]-2,2-dideuterio-acetate |
| InChI Formula | InChI=1 S/C6H9NO6/c8-4(9)1-7(2-5(10)11)3-6(12)13/h1-3H2,(H,8,9)(H,10,11)(H,12,13)/i1D2,2D2,3D2/hD3 |
| SMILES | [2 H]OC(=O)C([2 H])([2 H])N(C([2 H])([2 H])C(=O)O[2 H])C([2 H])([2 H])C(=O)O[2 H] |
| IUPAC Name | deuterio 2-[bis(1,1-dideuterio-2-deuteriooxy-2-oxoethyl)amino]-2,2-dideuterioacetate |
| Molecular Weight (g/mol) | 200.1942 |
| Formula Weight | 200.0995 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction