Learn More
Chemsplash Multi-Task Nitrile Powder Free Disposable Gloves, Style 4010
GREENER_CHOICE
Product Code. 30058274
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 1000
Product Code. 30058274
Brand: Chemsplash 4010.300.005
Only null left
Add to basket
Add to basket
Premium nitrile gloves with textured fingertips meet medical grade standards and are food contact approved.
  • Food Contact Approved
  • Medical Grade
  • Ambidextrous
  • Rolled Cuff and Latex Free
  • PFAS Free
  • Touchscreen Compatible
  • CE and UKCA Approved
  • Thickness: 2.8 mil, Weight: 3.5 g
  • AQL: 1.5
  • Suitable for chemical splash protection and have some elasticated properties, allowing for ease of use and fit.
  • BRC-approved work areas and the Pharmaceutical Industry.
  • Free from PFAS, RoHS, POP, TSCA, REACH, or any Conflict Materials.
Specifications
ANSI Abrasion Rating ANSI 0
Certifications/Compliance (Europe) EN374-1: KPT, EN374-5: VIRUS, EN16523, EN455: 1/2/3 Medical Devices, EN1186: Food Grade, EN420
Chlorinated No
Applications Laboratory
Color Blue
Finger Thickness (English) 2.8 mil
Finger Thickness (Metric) 0.07 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 2.8 mil
Palm Thickness (Metric) 0.07 mm
Product Type Disposable Gloves
Size Medium
Thickness (English) 2.8 mil
Thickness (Metric) 0.07 mm
EN 421 No
EN ISO 374-1 Type A
EN ISO 374 - 5 Bacteria, Fungi WITH Virus Protection
Glove Cuff Style Rolled
Lining Unlined