Learn More
Chemsplash Engi-Black Nitrile Powder Free Disposable Gloves, Style 4098
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 1000
Description
- Industrial Grade Nitrile Glove/Heavy Duty Nitrile
- Medical Grade and Food Contact Approved
- Micro Textured
- Ambidextrous
- Rolled Cuff and Latex Free
- PFAS Free
- Touchscreen Compatible
- CE and UKCA Approved
- Thickness: 4.3 mil, Weight: 5.0 g
- AQL: 1.5
- Nitrile gloves are designed for chemical splash protection and provide a comfortable fit thanks to their elasticated properties.
- Suitable for use in BRC approved work areas and the Pharmaceutical Industry.
- Product does not contain PFAS, RoHS, POP, TSCA, REACH, or any Conflict Materials.
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 0 |
| Certifications/Compliance (Europe) | EN374-1: KPT, EN374-5: VIRUS, EN16523, EN455: 1/2/3 Medical Devices, EN1186: Food Grade, EN420 |
| Chlorinated | No |
| Applications | Industrial |
| Color | Black |
| Finger Thickness (English) | 4.3 mil |
| Finger Thickness (Metric) | 0.11 mm |
| Latex Free | Yes |
| Material | Nitrile |
| Palm Thickness (English) | 4.3 mil |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur