Learn More
Hanna Instruments™ Nitrate Portable Photometer
Product Code. 11744476
Click to view available options
Range:
0 to 30mg/L
Unit Size:
Each
Product Code. 11744476
Brand: Hanna Instruments™ HI96728C
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Type Portable Photometer
Accuracy ±0.05mg/L
For Use With (Application) Nitrate
Height (Metric) 69 mm
Length (Metric) 192 mm
Light Source Tungsten Lamp
Range 0 to 30mg/L
Resolution 0.1mg/L
Weight (Metric) 0.36 kg
Width (Metric) 104 mm