missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NIH 3T3 Hypoxic/ Normoxic Cell Lysate

Product Code. 18620177 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18620177 2 Vials Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18620177 Supplier Novus Biologicals™ Supplier No. NBP311862
  • ONLINE EXCLUSIVE PRICE
    372.00€
    351.75€ / Pack of 2
    Save 20.25€
    5% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cell Lysate

TRUSTED_SUSTAINABILITY

Specifications

Preparation Method NIH 3T3 whole cell lysate is provided as a Western blot positive control. The lysates are provided as a set of treated and untreated samples. Use 10 μL / lane for a standard mini-gel blot (1.0 mg/mL).
Quantity 2 Vials
Host Species Mouse
Content And Storage Store at -20°C to -80°C. Avoid freeze/thaw cycles.
Target NIH 3T3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.