missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NIH 3T3 CoCl2 treated/Untreated Cell Lysate

Artikelnummer. 18674906 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Quantity:
2 Vials
Packungsgröße:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
18674906 2 Vials Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18674906 Lieferant Novus Biologicals™ Lieferanten-Nr. NBP311855
  • ONLINE EXCLUSIVE PRICE
    372.00€
    351.75€ / Pack of 2
    Save 20.25€
    5% Off
 Restricted Item
Estimated Shipment: 07-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Cell Lysate

TRUSTED_SUSTAINABILITY

Spezifikation

Preparation Method NIH 3T3 whole cell lysate is provided as a Western blot positive control. The lysates are provided as a set of treated and untreated samples. Use 10 μL / lane for a standard mini-gel blot (1.0 mg/mL).
Quantity 2 Vials
Host Species Mouse
Content And Storage Store at -20°C to -80°C. Avoid freeze/thaw cycles.
Target NIH 3T3
Name des Produkts
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.