Learn More
Nicotine Salicylate, TRC
Product Code. 30041310
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041310 25 mg 25mg
30005274 50 mg 50mg
30011957 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30041310 Supplier Toronto Research Chemicals Supplier No. TRCN42801025MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29790-52-1
Molecular Formula C10 H14 N2 . C7 H6 O3
Molecular Weight (g/mol) 300.35
Synonym 2-Hydroxybenzoic acid compd. with 3-[(2S)-1-methyl-2-pyrrolidinyl]pyridine (1:1), 2-Hydroxybenzoic acid compd. with (S)-3-(1-methyl-2-pyrrolidinyl)pyridine (1:1), Nicotine monosalicylate, (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine mono(2-hydroxybenzoate), 3-[(2S)-1-Methyl-2-pyrrolidinyl]pyridine mono(2-hydroxybenzoate), Salicylic acid compd. with nicotine (1:1), 1-Methyl-2-(3-pyridyl)pyrrolidine salicylate, Eudermol, Nicotine salicylate
IUPAC Name 2-hydroxybenzoic acid;3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
SMILES CN1CCC[C@H]1c2cccnc2.OC(=O)c3ccccc3O
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C10H14N2.C7H6O3/c1-12-7-3-5-10(12)9-4-2-6-11-8-9;8-6-4-2-1-3-5(6)7(9)10/h2,4,6,8,10H,3,5,7H2,1H3;1-4,8H,(H,9,10)/t10-;/m0./s1
Formula Weight 300.1474
Percent Purity >95%
Chemical Name or Material Nicotine Salicylate
Grade HPLC