Learn More
Nicotinamide adenine dinucleotide phosphate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 53-59-8 |
| Molecular Formula | C21H28N7O17P3 |
| MDL Number | MFCD00067537 |
| Quantity | 1 g |
| SMILES | O=C(C1=C[N+]([C@@H]2O[C@H](COP(OP(OC[C@H]3O[C@@H](N4C5=NC=NC(N)=C5N=C4)[C@H](OP(O)(O)=O)[C@@H]3O)(O)=O)([O-])=O)[C@@H](O)[C@H]2O)=CC=C1)N |
| Molecular Weight (g/mol) | 743.41 |
| Percent Purity | 98% |
| Chemical Name or Material | Nicotinamide adenine dinucleotide phosphate |