Learn More
Buerkle™ Nickle-Plated Brass Gastight Barrel Connector for Barrel Pump
Buerkle™ Nickle-Plated Brass Gastight Barrel Connector for Barrel Pump prevents hazardous gases from escaping from the barrel.
Brand: Buerkle™ 5601-0130
177.54 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Additional Details : Weight : 1.00350kg
Description
- The gas tight barrel screw joint prevents hazardous gases from escaping from the barrel
- R 2 in. steel barrel thread, non-return valve with connection 1/8 in.
- Nickel-plated brass, seals made of FKM
- The container connectors make decanting even easier and safer with their more secure connection to barrels
- The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange
Specifications
Barrel Screw Joint | |
Stainless Steel Barrel Pump | |
Non-return valve with connection 1/8 in. |
Nickel-Plated Brass, FKM Seals | |
Prevents hazardous gases from escaping |