Learn More
Usbeck Laborgeraete™ Nickel Plated Steel Forceps
Click to view available options
Length (Metric):
105 mm
130 mm
200 mm
300 mm
Unit Size:
Each
Specifications
Specifications
| Tip Style | Blunt |
| No. per Pack | 1 Pack |
| Material | Nickel-plated Steel |
| Length (Metric) | 200 mm |
| Dimensions (L x W) | 200 L mm |
| Type | Forceps |