Learn More
Usbeck Laborgeraete™ Nickel-plated Clamps with Vinyl-coated Jaws
Click to view available options
Diameter (Metric):
12 to 25 mm
30 to 80 mm
38 to 60 mm
Unit Size:
Each
Description
- Zinc diecasting
- Powdercoated
- Nickel plated vinyl coated jaws
Specifications
Specifications
| Quantity | 1 Ea. |
| Type | Clamp |
| Material | Nickel-plated |
| Diameter (Metric) | 30 to 80 mm |
| Coating | Vinyl |