missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel, Ni/Fe/Cr, Alloy HR-120, N08120, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Nickel, Ni/Fe/Cr, Alloy HR-120, N08120
Specifications
Specifications
| Chemical Name or Material | Aluminium, Antimony, Arsenic, Bismuth, Boron, Carbon, Chromium, Cobalt, Copper, Iron, Lead, Magnesium, Manganese, Molybdenum, Nickel, Niobium, Nitrogen, Oxygen, Phosphorus, Silicon, Silver, Tantalum, Tin, Titanium, Tungsten, Vanadium, Zirconium |
| Diameter (Metric) Outside | 38 mm |
| Form | Disc |
| Thickness (Metric) | 3 mm |
| Concentration or Composition (by Analyte or Components) | Oxygen: 0.0023%, Tin: 0.003%, Magnesium: 0.004%, Boron: 0.005%, Phosphorus: 0.016%, Titanium: 0.019%, Tungsten: 0.043%, Vanadium: 0.052%, Carbon: 0.054%, Copper: 0.0615%, Aluminium: 0.085%, Cobalt: 0.118%, Nitrogen: 0.207%, Molybdenum: 0.216%, Niobium: 0.57%, Silicon: 0.63%, Manganese: 0.81%, Chromium: 24.6%, Iron: 35.7%, Nickel: 37.1%, Bismuth: <0.0001%, Silver: 0.0001%, Antimony: 0.0004%, Lead: 0.001%, Zirconium: 0.002%, Arsenic: 0.003%, Tantalum: 0.005% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction