Learn More
Nickel, Ni/Cr/Mo/N, AL-6XN, N08367, ARMI∣MBH
Description
Nickel, Ni/Cr/Mo/N, AL-6XN, N08367, Ø38x20mm
Specifications
Specifications
| Chemical Name or Material | Carbon/Phosphorus/Aluminium/Tungsten/Vanadium/Cobalt/Niobium/Copper/Nitrogen/Silicon/Manganese/Molybdenum/Chromium/Nickel |
| CAS | 7440-02-0 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Carbon 0.0159 %/Phosphorus 0.0186 %/Aluminium 0.02 %/Tungsten 0.038 %/Vanadium 0.0513 %/Cobalt 0.0981 %/Niobium 0.15 %/Copper 0.187 %/Nitrogen 0.203 %/Silicon 0.275 %/Manganese 0.64 %/Molybdenum 6.16 %/Chromium 20.19 %/Nickel 23.89 % |