Learn More
Nickel Aluminum
Product Code. 11369147
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11369147
Brand: Thermo Scientific Alfa Aesar 039828.22
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Wear resistant coatings | CAS: 12635-27-7 | AlNi | 85.675 g/mol
Nickel Aluminum is used as wear resistant coatings. It is also used for the preparation of Raney™ Nickel catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 12635-27-7
Molecular Formula AlNi
Molecular Weight (g/mol) 85.675
MDL Number MFCD00167392
InChI Key NPXOKRUENSOPAO-UHFFFAOYSA-N
Synonym Aluminum Nickel
PubChem CID 6336846
IUPAC Name aluminum;nickel
SMILES [Al].[Ni]
Specifications
Color Black to Gray
Avg. Mol. Wt. or Mol. Wt. Range Ni:Al; 50:50 wt%
Quantity 100 g
Odor Odorless
Sensitivity Water reactive
Form Powder
UN Number UN3208
Solubility Information Insoluble in water.
Chemical Name or Material Nickel Aluminum
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Nickel Aluminum
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H228-Flammable solid.
  • H261-In contact with water releases flammable gases.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .