Learn More
Nickel, Ni/Fe/Cr, Alloy 20, N08020, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Nickel, Ni/Fe/Cr, Alloy 20, N08020, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Magnesium/Boron/Oxygen/Tin/Titanium/Aluminium/Phosphorus/Carbon/Nitrogen/Vanadium/Tungsten/Cobalt/Niobium/Silicon/Manganese/Molybdenum/Copper/Chromium/Nickel/Iron/Beryllium/Bismuth/Cerium/Indium/Palladium/Ruthenium/Selenium/Sodium/Tellurium/Antimony/Hafnium/Lanthanum/Rhenium/Silver/Yttrium/Germanium/Cadmium/Arsenic/Calcium/Tantalum/Zinc/Gallium/Zirconium/Lead |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Magnesium 0.0016 %/Boron 0.0036 %/Oxygen 0.005 %/Tin 0.008 %/Titanium 0.009 %/Aluminium 0.015 %/Phosphorus 0.019 %/Carbon 0.028 %/Nitrogen 0.034 %/Vanadium 0.089 %/Tungsten 0.13 %/Cobalt 0.14 %/Niobium 0.66 %/Silicon 0.7 %/Manganese 1.14 %/Molybdenum 2.1 %/Copper 3.46 %/Chromium 20 %/Nickel 32.6 %/Iron 38.7 %/Beryllium (<0.0001) %/Bismuth (<0.0001) %/Cerium (<0.0001) %/Indium (<0.0001) %/Palladium (<0.0001) %/Ruthenium (<0.0001) %/Selenium (<0.0003) %/Sodium (<0.0005) %/Tellurium (<0.0007) %/Antimony (<0.001) %/Hafnium (<0.001) %/Lanthanum (<0.001) %/Rhenium (<0.001) %/Silver (<0.001) %/Yttrium (<0.001) %/Germanium (<0.002) %/Cadmium (<0.005) %/Arsenic (<0.01) %/Calcium (<0.02) %/Tantalum (<0.02) %/Zinc (0.0013) %/Gallium (0.002) %/Zirconium (0.003) %/Lead (0.004) % |