missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NIBR LTSi

Product Code. 30226374
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30226374

Brand: Tocris Bioscience™ 8175/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective LATS inhibitor; activates YAP signaling

NIBR-LTSi is a potent and selective large tumor suppressor kinase (LATS) inhibitor (IC50 = 1.4 nM). Also activates YAP signaling (EC50 = 1.0 - 2.7 μM). Promotes liver organoid formation (EC50 = 0.35 μM), hepatocyte proliferation, ISC expansion, and blocks differentiation. NIBR-LTSi is orally bioavailable and suitable for use in vivo and in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-[(3R)-Piperidin-3-yl]oxyphenyl]-1H-pyrrolo[2,3-b]pyridin-4-amine
Quantity 50 mg
Target Other Kinase Inhibitors
Molecular Formula C18H20N4O
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.