missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NIBR LTSi
Description
NIBR-LTSi is a potent and selective large tumor suppressor kinase (LATS) inhibitor (IC50 = 1.4 nM). Also activates YAP signaling (EC50 = 1.0 - 2.7 μM). Promotes liver organoid formation (EC50 = 0.35 μM), hepatocyte proliferation, ISC expansion, and blocks differentiation. NIBR-LTSi is orally bioavailable and suitable for use in vivo and in vitro.
Specifications
Specifications
| Chemical Name or Material | N-[4-[(3R)-Piperidin-3-yl]oxyphenyl]-1H-pyrrolo[2,3-b]pyridin-4-amine |
| Quantity | 10 mg |
| Target | Other Kinase Inhibitors |
| Molecular Formula | C18H20N4O |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?