Learn More
Novus Biologicals™ NHSL2 293T Cell Transient Overexpression Lysate
Product Code. 18556983 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18556983 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18556983 Supplier Novus Biologicals™ Supplier No. H00340527T01
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target NHSL2
Synonym NHS-like 2, NHS-like protein 2
Gene Symbol NHSL2