Learn More
- Home
- Products
- Immunoassay Reagents and Kits
- Singleplex and Multiplex Bead Based Immunoassays
- R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit
R&D Systems NHP XL Cytokine Luminex™ Performance Premixed Kit
Product Code. 30002208
Change view
Click to view available options
Quantity:
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
5 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Each
39 product options available for selection
| Product Code. | Quantity | unitSize |
|---|---|---|
| 30002208 | 36 Plex | Each |
| 18717479 | 35 Plex | Each |
| 18792298 | 34 Plex | Each |
| 30002207 | 33 Plex | Each |
| 30002206 | 32 Plex | Each |
| 30002205 | 31 Plex | Each |
| 30002204 | 30 Plex | Each |
| 30002203 | 29 Plex | Each |
| 18700609 | 28 Plex | Each |
| 18724969 | 27 Plex | Each |
| 30002202 | 26 Plex | Each |
| 30002201 | 25 Plex | Each |
| 30002200 | 24 Plex | Each |
| 30002199 | 23 Plex | Each |
| 18742129 | 22 Plex | Each |
| 30002198 | 21 Plex | Each |
| 30002197 | 20 Plex | Each |
| 30002196 | 19 Plex | Each |
| 30002195 | 18 Plex | Each |
| 30002194 | 17 Plex | Each |
| 30002193 | 16 Plex | Each |
| 30002192 | 15 Plex | Each |
| 30002191 | 14 Plex | Each |
| 18703619 | 13 Plex | Each |
| 18729578 | 12 Plex | Each |
| 30002190 | 11 Plex | Each |
| 30002189 | 10 Plex | Each |
| 18795189 | 9 Plex | Each |
| 30002188 | 7 Plex | Each |
| 30002187 | 6 Plex | Each |
| 18707479 | 5 Plex | Each |
| 18709787 | 4 Plex | Each |
| 30002186 | 3 Plex | Each |
| 30002185 | 2 Plex | Each |
| 18441820 | 40 Plex | 1 set |
| 18451820 | 38 Plex | 1 set |
| 18422530 | 39 Plex | 1 set |
| 18452080 | 37 Plex | 1 set |
| 18411410 | 8 Plex | 1 set |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 39 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Quantity | 36 Plex |
| Species | Primate |
| Binding Target | Multiplex Cytokine Assay Kits |
| For Use With (Application) | ELISA |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |