Learn More
R&D Systems NHP XL CXCL2/GRO beta, Magnetic Luminex™ Performance Assay
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Primate |
| Binding Target | CXCL2/GRO beta/MIP-2/CINC-3 |
| Gene ID (Entrez) | 2920 |
| Synonym | chemokine (C-X-C motif) ligand 2, CINC-2a, CINC3, CINC-3, C-X-C Motif Chemokine 2, GRO beta, GRO2, GRO2 oncogene, GROb, Gro-beta, Growth-regulated protein beta, Macrophage inflammatory protein 2-alpha, melanoma growth stimulatory activity beta, MGSA beta, MGSA-b, MGSA-beta, MIP2, MIP-2, MIP-2a, MIP2A, MIP2-alpha, SCYB2 |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |