Learn More
Abnova™ NFIB/TCF3 SY Translocation FISH Probe
Product Code. p-7190350
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Product Code. 16083555
Brand: Abnova™ FT0014.100uL
Labeled FISH probes for identification of gene transloaction using Fluoresecent In Situ Hybridization Technique.
Specifications
Content And Storage Store at 4°C in the dark.
Description DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Format Tube
For Use With (Application) FISH-Ce
Gene ID (Entrez) 4781/6929
Includes DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Quantity 100 μL
Form Liquid