Learn More
Thermo Scientific New Equipment Startup Service for Cold Storage, Freezer, Refrigerator, ULT AND Arctiko CRYO -150°C Compressor-based Freezer
Artikelnummer. 32176308 Alle ansehen Thermo Scientific Produkte
Ansicht ändern
Click to view available options
For Use With (Equipment):
ULT Freezers
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. For Use With (Equipment) Packungsgröße
32176308 ULT Freezers Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 32176308 Lieferant Thermo Scientific Lieferanten-Nr. STARTULTMVS
 Restricted Item
Estimated Shipment: 07-01-2027
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Installation service scope covers compressor-based refrigerators, freezers, ULT and Arctiko CRYO -150°C equipment where LN2 backup is secondary to the primary cooling system.
  • Not suitable for liquid nitrogen Dewar-type cryopreservation systems
  • White-glove delivery, building works and primary LN2 cryopreservation systems are excluded unless separately quoted
TRUSTED_SUSTAINABILITY
Spezifikation
For Use With (Equipment) ULT Freezers
Product Type Service
Name des Produkts
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.