Learn More
Eppendorf™ New Brunswick™ Innova™ 43, 230 V, orbit 5.1 cm (2 in)
Product Code. 11300025
Change view
Click to view available options
Orbit:
51 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Orbit Unit Size
11300025 51 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 11300025 Supplier Eppendorf™ Supplier No. M13200012
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
The Eppendorf Innova 43 and 43R (refrigerated) console incubator shakers are the ideal solution for precise handling of large culture volumes. The pedal operated, top loading design allows the easy vertical placement of flasks, and samples can be viewed through the window.
New Brunswick™ Innova™ 43, 230 V, orbit 5.1 cm (2 in)- Console style shaker/incubator with triple-eccentric drive – provides large capacity shaking for flasks up to 6 L - Continuous mode or multiple-step programming- Programmable Innova controls – automatically changes temperature, speed, and optional photosynthetic and UV germicidal lights at timed intervals (Store up to 4 individual programs with 15 steps each)- Top mounted controls and display – for easy, ergonomic viewing and setting - Foot pedal release with easy to opening lid – provides hands-free door opening/closing operation and effortless access to samples- Built in spill pan catches accidental spills to aid cleaning and protects drive mechanism. Also allows use as a water reservoir to reduce sample evaporation- Large 76 × 46 cm (30 in.× 18 in.) platform size provides increased flexability with the number and size of cultures (sold separately)
TRUSTED_SUSTAINABILITY
Specifications
Orbit 51 mm
Speed Range 25 to 500 rpm with 2.5 cm orbit, 25 to 300 rpm with 5.1 cm orbit
Electrical Requirements 230 V, 50 Hz
Voltage 230 V
Temperature Range (Metric) Ambient +5° to 80°C
Frequency 50 Hz
Depth (English) 20 in.
Depth (Metric) 81.3 cm
Dimensions (L x W x H) 81.3 x 115.6 x 103.1 cm (45.5 x 20 x 40.6 in.)
Height (English) 40.6 in.
Width (English) 45.5 in.
Width (Metric) 115.6 cm
Timer Range 0.01 to 99:59 hr., Continuous