Learn More
Eppendorf™ Scientific shaker Microplate Platform
Product Code. 11309854
Change view
Click to view available options
No. of Positions:
5
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Positions Unit Size
11309854 5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11309854 Supplier Eppendorf™ Supplier No. TTR221
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
Estimated Shipment: 09-12-2026
For use with New Brunswick Innova™, I-Series™ and Excella™ Shakers
Mount on universal platforms
Capacities listed are the most vessels of each size that will fit in a balanced pattern.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Microplate Rack
For Use With (Equipment) Innova 2100, 2150, 2300, 2350; Excella E10, E24/E24R, E25/E25R; all I-Series
No. of Positions 5