Learn More
Eppendorf™ I-Series Stackable Shakers: Stacking Kits
Description
Requires
Sticky Pad Platform (14-285-746) requires Sticky Pad (14-285-753, sold separately)
- Stacking kit enables two I-26 Series Stackable Shakers to be stacked vertically
- Bases allow shaker(s) to be raised to suitable height
Specifications
Specifications
| Product Type | Base Riser for Single Stack |
| Height (English) | 17 in. |
| Height (Metric) | 43.18 cm |
| For Use With (Equipment) | NBS I-Series Stackable Shakers |
| Size | Tall |