Learn More
Eppendorf™ Gable Covers for Model C76 Water Bath Shaker
Click to view available options
Material:
Plexiglass™
Unit Size:
Each
Description
- Plexiglas™ cover permits temperatures up to 60°C Stainless-steel cover required for temperatures above 60°C
Specifications
Specifications
| For Use With (Equipment) | Model C76 water bath shaker |
| Material | Plexiglass™ |