Learn More
Eppendorf™ Gable Covers for Model C76 Water Bath Shaker
Product Code. 11379874
Click to view available options
Material:
Plexiglass™
Unit Size:
Each
Product Code. 11379874
Brand: Eppendorf™ M12312010
Only null left
Add to basket
Add to basket
For C76 Water Bath Shaker
  • Plexiglas™ cover permits temperatures up to 60°C Stainless-steel cover required for temperatures above 60°C
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Model C76 water bath shaker
Material Plexiglass™