Learn More
Decon™ Neutracon Liquid Detergent
Product Code. 11721178
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
Product Code. 11721178
Brand: Decon™ NEU5
Mild detergent for fragile instruments. Neutracon™ Liquid Detergent is suitable for washing or decontamination of all non-ferrous metals (aluminum, zinc, copper, silver, brass), polymers and any materials sensitive to acidic or alkaline detergents.
  • Biodegradable
  • Phosphate-free
  • Non-corrosive
  • Non-toxic
  • Fully rinsable
  • Non-flammable
  • Diluted in water by 1 to 3%
  • Reduced time in ultrasonic cleaner with heating up to 50 ° C
Specifications
Product Type Liquid Detergent
Form Liquid
Volume (Metric) 5 L