missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Neuropeptide Y, Porcine
Click to view available options
Quantity:
0.5 mg
1 mg
missing translation for 'unitSize'
0.5mg
1mg
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Species | Porcine |
| Purification Method | HPLC |
| Content And Storage | -30°C to -10°C |
| Format | Powder |
| Molecular Weight (g/mol) | 4253.70 |
| Product Type | Neuropeptide Y, porcine |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Sequence | NPY |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur