Learn More
STEMCELL Technologies NeuroFluor™ NeuO
Product Code. 17199991
Encompass_Preferred
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Product Code. 17199991
Brand: STEMCELL Technologies 01801
Membrane-permeable fluorescent probe for the detection of live neurons
  • NeuroFluor™ NeuO is a membrane-permeable fluorescent probe that selectively labels primary and pluripotent stem cell-derived neurons in live cultures (J C Er et al.) Angew Chem Int Ed, 2015)
  • Cells labeled with NeuroFluor™ NeuO can be visualized using fluorescent imaging
  • Labeling with this probe is non-permanent; it can be washed off, providing unlabeled, viable cells for downstream applications
  • For additional information, see References
Specifications
Description Neural Cells, PSC-Derived; Neurons
For Use With (Application) Characterization; Phenotyping
Quantity 0.1 mL
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.