missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies NeuroCult™ SM1 Neuronal Supplement
Description
- Avoid culture failure with a Brewer's B27-based supplement that provides a consistent neuronal culture experience for CNS-derived or human pluripotent stem cell (hPSC)-derived cells
- NeuroCult SM1 (STEMCELL Modified-1) Neuronal Supplement is based on the published formulation (Brewer et al. Brain Res, 1989) and standardized to more reproducibly support survival and maturation of functional neurons
- Cultures with this serum-free supplement feature increased neurite outgrowth and branching in short- and long-term cultures
- NeuroCult SM1 may also be used as a serum-replacement supplement for various customizable applications, such as neurotoxicity assays and calcium imaging
- For your convenience, NeuroCult SM1 is included as a component of multiple BrainPhys Neuronal Medium culture kits for primary and hPSC-derived neurons (Catalog #05792, 05793, 05794, and 05795)
Specifications
Specifications
| Cell Type | Neural Cells, PSC-Derived, Neurons, Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Differentiation, Maintenance |
| Quantity | 10 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?