missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies NeuroCult™ Proliferation Supplement (Mouse & Rat)
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
Description
- NeuroCult™ Proliferation Supplement (Mouse & Rat) is a standardized, serum-free supplement for the culture of mouse and rat neural stem and progenitor cells
- NeuroCult™ Proliferation Supplement (Mouse & Rat) is designed to be combined with NeuroCult™ Basal Medium (Mouse & Rat; Catalog #05700) and appropriate cytokines (not included)
- NeuroCult™ Proliferation Supplement (Mouse & Rat) is optimized to maintain mouse and rat neural stem cells in culture for extended periods of time without the loss of their self-renewal, proliferation, or differentiation potential
- This supplement is also a component of NeuroCult™ Proliferation Kit (Mouse & Rat; Catalog #05702)
- NOTE: When preparing Complete NeuroCult™ Proliferation Medium, addition of Human Recombinant EGF (Catalog #78006.1) is required
- When culturing cells obtained from adult mouse or rat, Human Recombinant bFGF (Catalog #78003.1) and Heparin Solution (Catalog #07980) are also required
Specifications
Specifications
| Cell Type | Brain Tumor Stem Cells, Neural Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Expansion, Functional Assay, Spheroid Culture |
| Quantity | 50 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction