missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies NeuroCult™ NS-A Proliferation Kit (Human)
Description
- NeuroCult™ NS-A Proliferation Kit (Human) is a standardized, serum-free basal medium and supplement for the culture of human neural stem and progenitor cells from normal tissues or tumor samples, in the neurosphere or adherent monolayer system
- When supplemented with appropriate cytokines, NeuroCult™ NS-A Proliferation Kit (Human) is optimized to maintain human neural stem cells in culture for extended periods of time without the loss of their self-renewal, proliferation, or differentiation potential
- NOTE: Addition of rh EGF (Catalog #78006), rh bFGF (Catalog #78003) and heparin (Catalog #07980) is required
Specifications
Specifications
| Cell Type | Brain Tumor Stem Cells, Neural Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Expansion, Functional Assay, Spheroid Culture, Toxicity Assay |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?