missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Neuro2a Chloroquine Treated/ Untreated Cell Lysate

Product Code. 18675296 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 Vials
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18675296 2 Vials Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18675296 Supplier Novus Biologicals™ Supplier No. NBP249688
  • ONLINE EXCLUSIVE PRICE
    207.00€
    195.30€ / Pack of 2
    Save 11.70€
    5% Off
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cell Lysate

TRUSTED_SUSTAINABILITY

Specifications

Preparation Method Neuro2a Chloroquine treated/ untreated lysates are provided as positive and negative control for Western blot analysis in Autophagy research. The lysates are provided as one vial of treated and one vial of untreated samples. Use 10 μL / lane for a standard mini-gel blot (approx. 1 mg/mL).
Quantity 2 Vials
Content And Storage Aliquot and store at -20°C to +80°C. Avoid freeze/thaw cycles.
Research Category Autophagy, Cellular Markers, Chaperone Mediated Autophagy (CMA), mTOR Pathway, Signal Transduction, Ubiquitin Proteasome Pathway
Target Neuro2a
Synonym LC3 Lysate, lc3b autophagy marker, LC3B western blot
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.