Learn More
KNF™ Neuberger Diaphragm Vacuum Pump N 920 G with EU-Plug
Product Code. 15720868
Click to view available options
Flow Rate:
21 L/min.
Unit Size:
Each
Product Code. 15720868
Brand: KNF™ 302473/057335
Only null left
Add to basket
Add to basket
Chemically resistant, oil-free diaphragm pump with integrated speed control for adjusting the pump speed to your specific process requirements. It features a gas ballast, PPS heads, PTFE-coated diaphragms and FFPM valves.
  • Ultimate Vacuum (max.): 2 mbar (abs.)
  • Adjustable speed control
  • High suction speed, particularly in the low vacuum range
  • Ideal for extremely aggressive/corrosive gases and vapors
  • Clean, 100% oil-free operation
Specifications
Max. Vacuum 2 Mbar abs.
Motor Type AC Motor
Dimensions (L x W x H) 324 x 158 x 226 mm
Description Diaphragm Vacuum Pump
Material Diaphragm: PTFE-coated, Pump Head: PPS, Valves: FFPM
Model Diaphragm Vacuum Pump N 920 G
Plug Type EU
Wattage 135 W
Weight (English) 9,5 kg
Flow Rate 21 L/min.
Frequency 50/60 Hz
Overall Height (English) 226 mm
Overall Length (English) 324 mm
Overall Width (English) 158 mm
Pump Type Vacuum System
Enclosure Rating IP20
No. of Pump Heads 3 heads
Oil Capacity Oil free
Applications Rotary Evaporation, Distillation, Centrifugal Concentration, Degassing, Dessiccation, Gel Drying
No. of Stages 2 stages
Maximum Pressure 0.5 bar G
Voltage 100/240 V
Quantity 1