missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nervonic-16,17,17,18,18,19,19,20,20,21,21,22,22,23,23,24,24,24-d18 Acid, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Nervonic Acid-16,17,17,18,18,19,19,20,20,21,21,22,22,23,23,24,24,24-D18 |
| Molecular Formula | C24 D18 H28 O2 |
| Quantity | 0.005 g |
| Synonym | (15 Z)-15-Tetracosenoic-16,17,17,18,18,19,19,20,20,21,21,22,22,23,23,24,24,24-d18 acid, Nervonic acid-d18, Nervonic-16,17,17,18,18,19,19,20,20,21,21,22,22,23,23,24,24,24-d18 Acid, Nervonic Acid-16,17,17,18,18,19,19,20,20,21,21,22,22,23,23,24,24,24-D18 |
| InChI Formula | InChI=1 S/C24H46O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24(25)26/h9-10 H,2-8,11-23H2,1H3,(H,25,26)/b10-9-/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9 D |
| SMILES | [2 H]\C(=C\CCCCCCCCCCCCCC(=O)O)\C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | (Z)-16,17,17,18,18,19,19,20,20,21,21,22,22,23,23,24,24,24-octadecadeuteriotetracos-15-enoic acid |
| Molecular Weight (g/mol) | 384.7318 |
| Formula Weight | 384.4628 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction