Learn More
Nepsilon-Acetyl-Nalpha-Boc-L-lysine, 98%, Thermo Scientific™
Product Code. 15488868
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15488868
Brand: Thermo Scientific Alfa Aesar H62347.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6404-26-8
Molecular Formula C13H24N2O5
Molecular Weight (g/mol) 288.34
MDL Number MFCD00057791
InChI Key IOKOUUAPSRCSNT-UEQNJFAPNA-N
Synonym boc-lys ac-oh, n-boc-n'-acetyl-l-lysine, s-6-acetamido-2-tert-butoxycarbonyl amino hexanoic acid, boc-lys-ac-oh, n-boc-n-acetyl-l-lisine, boc-n-epsilon-acetyl-l-lysine, na-t-boc-n-epsilon-acetyl-l-lysine, 2s-6-acetylamino-2-tert-butoxycarbonylamino hexanoic acid, 2s-2-tert-butoxycarbonyl amino-6-acetamidohexanoic acid
PubChem CID 7016049
IUPAC Name (2S)-6-acetamido-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O
Specifications
Quantity 1 g
Formula Weight 288.34
Percent Purity 98%
Chemical Name or Material Nepsilon-Acetyl-Nalpha-Boc-L-lysine