Learn More
Nemonapride, TRC
Specifications
Specifications
| Chemical Name or Material | Nemonapride |
| CAS | 75272-39-8 |
| Molecular Formula | C21 H26 Cl N3 O2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C21H26ClN3O2/c1-14-18(9-10-25(14)13-15-7-5-4-6-8-15)24-21(26)16-11-17(22)19(23-2)12-20(16)27-3/h4-8,11-12,14,18,23H,9-10,13H2,1-3H3,(H,24,26)/t14-,18-/m1/s1 |
| SMILES | CNc1cc(OC)c(cc1Cl)C(=O)N[C@@H]2CCN(Cc3ccccc3)[C@@H]2C |
| IUPAC Name | N-[(2R,3R)-1-benzyl-2-methylpyrrolidin-3-yl]-5-chloro-2-methoxy-4-(methylamino)benzamide |
| Molecular Weight (g/mol) | 387.9 |
| Formula Weight | 387.1714 |