Learn More
Thermo Scientific™ Neisseria Selective Agar
Product Code. 13225349
Click to view available options
Description:
Neisseria Selective Agar
Quantity:
10/Pk.
Unit Size:
Pack of 10
Product Code. 13225349
Brand: Thermo Scientific™ PO5004A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Isolate and cultivate pathogenic Neisseria species with a special formulation, including a peptone mixture of meat and plant enzymatic digests, starch and phosphate buffers.
Isolate and cultivate pathogenic Neisseria species with Thermo Scientific™ Neisseria Selective Agar, a medium specially formulated with a peptone mixture that includes meat and plant enzymatic digests and the presence of starch to ensure that toxic metabolites produced by Neisseria are absorbed. Phosphate buffers are also included to prevent changes in pH due to amine production that would affect the survival of the organism.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Tested in accordance with CEN ISO/TS 11133-2. Formulation of this medium conforms to ISO 4833.
Color Signal brown, opaque
Content And Storage 6°C to 12°C
Description Neisseria Selective Agar
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.