Learn More
Aalborg™ Needle Valve for Brass Flowmeter
Click to view available options
Airflow Rate:
20000 mL/min.
5000 mL/min.
60000 mL/min.
Waterflow Rate:
1200mL/min.
3500mL/min.
350mL/min.
Unit Size:
Each
Description
- Available in three conveniently overlapping orifice-needle sizes.
- These versatile, rugged and reliable, valves are suitable for laboratory instrumentation, bench top or OEM flow control purposes.
- Valves are offered in straight (T) and 90 degree (L) flow patterns. All valves are supplied with 1/8in. FNPT inlet and outlet ports.
- Valve cartridges are also interchangeable with built-in valves of Aalborg’s series of P, T, S, and G flow meter product line.
- Bubble tight shutoff
Specifications
Specifications
| For Use With (Equipment) | 150 mm Gas/Liquid Flowmeter |
| Type | Standard Valve |
| Airflow Rate | 20000 mL/min. |
| Waterflow Rate | 1200mL/min. |
| Max. Operating Temperature (Metric) | 82°C |